Skip to Content
Merck
All Photos(1)

Key Documents

1376505

USP

Mechlorethamine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Mechlorethamine hydrochloride, 2-Chloro-N-(2-chloroethyl)-N-methylethanamine, N-Methylbis(2-chloroethyl)amine hydrochloride, Bis(2-Chloroethyl)methylamine, Bis(2-chloroethyl)methylamine, MBA hydrochloride, NM, Nitrogen mustard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CH2)2NCH3·HCl
CAS Number:
Molecular Weight:
192.51
MDL number:

manufacturer/tradename

USP

mp

108-111 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

Cl.CN(CCCl)CCCl

InChI

1S/C5H11Cl2N.ClH/c1-8(4-2-6)5-3-7;/h2-5H2,1H3;1H

InChI key

QZIQJVCYUQZDIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Carc. 1B - Muta. 1B - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service