Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2986

Sigma-Aldrich

Anti-PU.1/Spi1 Antibody, clone 1B8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

31 kDa-transforming protein, SPI1, Transcription factor PU.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B8, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 31.08 kDa
observed mol wt ~36 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

bovine, porcine

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SPI1(6688)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Transcription factor PU.1. It targets an epitope within 22 amino acids from the N-terminal half of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal half of human Transcription factor PU.1 (SPI1).

Application

Quality Control Testing

Evaluated by Western Blotting in Daudi cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Transcription factor PU.1 (SPI1) in Daudi cell lysate.

Tested Applications

Affinity Binding Assay: The binding affinity of this antibody to PU.1-Spi1 peptide (100 g/ml) was at least one thousand-fold (1,000X) higher than that obtained with the non-specific control peptide.

Immunoprecipitation Analysis: 2 µg from a representative lot immunoprecipitated PU.1/Spi1 in Daudi cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PU.1/Spi1 in HL-60 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Transcription factor PU.1 (UniProt: P17947; also known as 31 kDa-transforming protein) is encoded by the SPI1 gene (Gene ID: 6688) in human. SPI1 gene has been mapped to human chromosome 11p11.2. It belongs to the E26 transformation specific (Ets) family of transcription factors that binds (via the ETS domain) onto the purine-rich DNA core sequence 5′-GAGGAA-3′, also known as the PU-box. It can also can bind to RNA and interfere with pre-mRNA splicing. PU.1 has a N-terminal transcriptional activation domain with acidic amino acid sub domains, a central proline, glutamic acid, serine, and threonine-rich (PEST) region and a C-terminal (ETS) domain. The protein is mainly expressed in cells of the hematopoietic lineage. PU.1 is reported to be essential for the differentiation of myeloid cells and serves as a transcription activator involved in macrophage proliferation. PU.1 is overexpressed in some B cell lymphoma, Hodgkin, and non- Hodgkin lymphoma. Along with other transcription factors, it plays a vital role in fetal hematopoiesis and controls cellular communication in the immune system. High expression of Spi1 leads to senescence in erythroid progenitors and is regarded as critical step to target leukemia. Deletions in the SPI1 gene is implicated in acute myeloid leukemia (AML). SPI1 gene inactivation results in fetal death. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service