ZRB2902
Anti-CHD2 Antibody, clone 2J13 ZooMAb® Rabbit Monoclonal


recombinant, expressed in HEK 293 cells
Synonym(s):
ATP-dependent helicase CHD2, Chromodomain-helicase-DNA-binding protein 2
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2J13, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 211.34 kDa
observed mol wt ~211 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
rat, mouse
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... CHD2(1106)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HaCaT cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected CHD2 in HaCaT cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CHD2 in lysate from HEK-293 cells nuclear extract.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CHD2 in SH-SY5Y cells.
Affinity Binding Assay: A representative lot of this antibody bound with CHD2 peptide at least one thousand-fold (1,000X) higher affinity than with non-specific control peptide.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CHD2 in human cerebral cortex tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service