SRP5027
EPHA4 (610-887), active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
HEK8, SEK, TYRO1
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
70-95 nmol/min·mg
mol wt
~58 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EPHA4(2043)
General description
EPHA4 also known as EPH receptor A4, belongs to the ephrin receptor subfamily of protein-tyrosine kinases which have been implicated in mediating developmental events, particularly in the nervous system. The EPHA4 ligand ephrin-A3 is localized to the astrocytic processes that envelop the spine. Activation of EPHA4 by ephrin-A3 induces spinal retraction and reduces spine density and inhibits the interaction distorted spine shape and organization. EPHA4-null mice possess defects in the corticospinal tract and anterior commissure indicating a model in which an ephrin ligand on the axons senses EPHA4 on spinal cord cells surrounding the corticospinal tract.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Dottori et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(22), 13248-13253 (1998-10-28)
Members of the Eph family of tyrosine kinase receptors have been implicated in the regulation of developmental processes and, in particular, axon guidance in the developing nervous system. The function of the EphA4 (Sek1) receptor was explored through creation of
J G Flanagan et al.
Annual review of neuroscience, 21, 309-345 (1998-04-08)
The Eph receptors are the largest known family of receptor tyrosine kinases. Initially all of them were identified as orphan receptors without known ligands, and their specific functions were not well understood. During the past few years, a corresponding family
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service