SRP0347
EphB4 active human
recombinant, expressed in baculovirus infected Sf9 cells
Synonym(s):
EPH receptor B4, HTK, MYK1, TYRO11
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
form
aqueous solution
mol wt
50 kDa
packaging
pkg of 10 μg
technique(s)
activity assay: suitable
inhibition assay: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EPHB4(2050)
General description
Human EPHB4 ( GenBank Accession No. NM_004444), amino acids 563-987 with N-terminal His tag, MW=50 kDa, expressed in a Baculovirus infected Sf9 cell expression system.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Unit Definition
One unit is defined as the amount of enzyme that will transfer 1 nmol phosphate to tyrosine substrate per minute at pH 7.4 and 30° C.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thomas Bruhl et al.
Circulation research, 94(6), 743-751 (2004-02-07)
Homeobox genes (Hox) encode for transcription factors, which regulate cell proliferation and migration and play an important role in the development of the cardiovascular system during embryogenesis. In this study, we investigated the role of HoxA9 for endothelial cell migration
Yusuke Takahashi et al.
Anticancer research, 34(6), 2913-2918 (2014-06-13)
Signaling induced by binding of erythropoietin-producing hepatoma-amplified sequence (EPH) receptors to their cell-surface ephrin ligands is implicated in hematopoiesis and growth of various cancer cells. However, the roles of EPH-ephrin signaling in leukemia have not been elucidated. We investigated the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service