SRP0325
PHF8 human
recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)
Synonym(s):
JHDM1F, MRXSSD, PHD finger protein 8, ZNF422
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥80% (SDS-PAGE)
form
aqueous solution
mol wt
52 kDa
packaging
pkg of 20 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PHF8(23133)
General description
Human PHF8 (also known as JHDM1F), (GenBank Accession No. NM_001184896), amino acids 2 - 450 with N-terminal FLAG-tag, MW=52 kDa, expressed in a Baculovirus infected Sf9 cell expression system.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Physical form
Formulated in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 80 ug/mL Flag peptide, 20% Glycerol, and 3 mM DTT.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wen Liu et al.
Nature, 466(7305), 508-512 (2010-07-14)
While reversible histone modifications are linked to an ever-expanding range of biological functions, the demethylases for histone H4 lysine 20 and their potential regulatory roles remain unknown. Here we report that the PHD and Jumonji C (JmjC) domain-containing protein, PHF8
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service