SRP0127
DNMT2 Active human
recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)
Synonym(s):
DNA (cytosine-5-)-methyltransferase 2, PUMET, RNMT1, tRNA (cytosine-5-)-methyltransferas, tRNA aspartic acid methyltransferase 1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
human
recombinant
expressed in baculovirus infected insect cells
Assay
≥80% (SDS-PAGE)
form
aqueous solution
mol wt
71 kDa
packaging
pkg of 10 μg
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... TRDMT1(1787)
General description
DNA (cytosine-5-)-methyltransferase 2 (DNMT2) is a highly conserved protein which is part of the DNA methyltransferase family. It possesses a conserved cysteine residue in its catalytic pocket. The gene encoding it is localized on human chromosome 10p14->p12.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Biochem/physiol Actions
DNA (cytosine-5-)-methyltransferase 2 (DNMT2) methylates the cytosine-38 residue of the aspartic acid transfer RNA (tRNA-Asp). It has been shown to interact with the anticodon stem and loop. The protein also functions in cellular physiology and stress response. It is significantly expressed in cancers.
Physical form
Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20 and 10% glycerol.
Preparation Note
Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Azacytidine Inhibits RNA Methylation at DNMT2 Target Sites in Human Cancer Cell Lines
Matthias Schaefer
Cancer Research, 69(20) (2009)
Human DNMT2 methylates tRNAAsp molecules using a DNA methyltransferase-like catalytic mechanism
Tomasz P. Jurkowski
RNA, 14(8), 1663-1670 (2008)
Assignment1 of candidate DNA methyltransferase gene (DNMT2) to human chromosome band 10p15.1 by in situ hybridization
Vilain A
Cytogenetic and genome research, 82 (1998)
Mapping the tRNA binding site on the surface of human DNMT2 methyltransferase.
Jurkowski TP
Biochemistry, 51(22), 4438-4444 (2012)
Winfried Elhardt et al.
Biochimie, 112, 66-72 (2015-03-10)
Methylation of tRNA is an important post-transcriptional modification and aberrations in tRNA modification has been implicated in cancer. The DNMT2 protein methylates C38 of tRNA-Asp and it has a role in cellular physiology and stress response and its expression levels
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service