Skip to Content
Merck
All Photos(1)

Key Documents

SML4126

Ginsenoside Rd

new

≥98% (HPLC)

Synonym(s):

(3β,12β)-20-(β-D-Glucopyranosyloxy)-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, 3-O-β-D-Glucopyranosyl-(1→2)-β-D-glucopyranosyl-20-O-β-D-glucopyranosyldammar-24-ene-3β,12β,20S-triol, Chikusetsusaponin FK7, Ginsenoside Rd1, Gypenoside VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 17 to 23°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Triterpene saponin isolated from ginseng that demonstrates a range of pharmacological effects, such as anti-inflammatory, antioxidant, anti-cancer, neuroprotective, and cardiovascular protective properties.

Ginsenoside Rd, a potent triterpene saponin, derived from ginseng demonstrates a range of pharmacological effects, such as anti-inflammatory, antioxidant, anti-cancer, neuroprotective, and cardiovascular protective properties. Potential advantages include enhancing cognitive function, diminishing oxidative stress, and regulating the immune system. Ginsenoside-Rd acts on diverse signaling pathways, including the NF-κB pathway associated with inflammation. Moreover, Ginsenoside Rd is implicated in influencing neuronal function and shielding the brain from oxidative stress.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service