Skip to Content
Merck
All Photos(1)

Key Documents

SML2724

Sigma-Aldrich

CC2-49

≥98% (HPLC)

Synonym(s):

EN6 inactive control, EN6 negative control, N-(3-Acetamido-4-fluorophenyl)-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide, N-[3-(Acetylamino)-4-fluorophenyl]-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14F2N4O2
CAS Number:
Molecular Weight:
356.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(cccc1)[n]2ncc(c2)C(=O)Nc3cc(c(cc3)F)NC(=O)C

InChI

1S/C18H14F2N4O2/c1-11(25)22-16-8-13(6-7-14(16)19)23-18(26)12-9-21-24(10-12)17-5-3-2-4-15(17)20/h2-10H,1H3,(H,22,25)(H,23,26)

InChI key

NXUJHBGKNXUNKK-UHFFFAOYSA-N

Biochem/physiol Actions

CC2-49 is a cysteine non-reactive EN6 analog that lacks lysosomal vacuolar H(+)-ATPase (v-ATPase)-modifying activity, cellular mTORC1 inhibitory potency, nor autophagy-inducing efficacy. CC2-49 is the recommended negative (inactive) control for EN6.
Cysteine non-reactive analog of and the recommended negative (inactive) control for EN6.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service