SAB5701979
Anti-GTF2F2 Antibody, clone 3T7Q9, Rabbit Monoclonal
Synonym(s):
GTF2F2, BTF4, RAP30, TF2F2, TFIIF, general transcription factor IIF subunit 2
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
3T7Q9, monoclonal
form
liquid
mol wt
30 kDa
species reactivity
rat, rat, human
concentration
1.50 mg/mL
technique(s)
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
MAERGELDLTGAKQNTGVWLVKVPKYLSQQWAKASGRGEVGKLRIAKTQGRTEVSFTLNEDLANIHDIGGKPASVSAPREHPFVLQSVGGQTLTVFTESS
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... GTF2F2(2963)
General description
Immunogen
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service