SAB4702032
Monoclonal Anti-CD120a purified low endotoxin produced in mouse

Synonym(s):
FPF, TBP1, TNF R1, TNFR55, TNFRSF1A, p60
conjugate
unconjugated
Quality Level
antibody product type
primary antibodies
clone
H398, monoclonal
form
buffered aqueous solution
species reactivity
human
concentration
1 mg/ml
technique(s)
FUNC: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2a
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... TNFRSF1A(7132)
General description
Specificity
Immunogen
Physical form
Preparation Note
Storage and Stability
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service