Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500962

Sigma-Aldrich

Anti-NPAS3 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPAS3(64067)

Immunogen

Antibody was raised against a 28 amino acid synthetic peptide from near the center of human NPAS3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500962.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric W Brunskill et al.
The European journal of neuroscience, 22(6), 1265-1276 (2005-09-30)
Npas3 is a member of the bHLH-PAS superfamily of transcription factors that is expressed broadly in the developing neuroepithelium. To study the function of this gene, mice deficient in Npas3 were generated and characterized. Npas3-/- mice were growth-retarded and exhibited
E W Brunskill et al.
Mechanisms of development, 88(2), 237-241 (1999-10-27)
Here we describe the cloning and expression pattern of a new bHLH-PAS domain gene, Npas3. Npas3 shares 50.2% amino acid sequence identity with Npas1 and a lesser similarity with other members of the bHLH-PAS domain family of transcription factors. Northern
D Kamnasaran et al.
Journal of medical genetics, 40(5), 325-332 (2003-05-15)
Schizophrenia and its subtypes are part of a complex brain disorder with multiple postulated aetiologies. There is evidence that this common disease is genetically heterogeneous, with many loci involved. In this report, we describe a mother and daughter affected with

Global Trade Item Number

SKUGTIN
SBP3500962-0.05MG4061832174129
SAB3500962-100UG4061832497778

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service