Skip to Content
Merck
All Photos(1)

Key Documents

SAB2108690

Sigma-Aldrich

Anti-MNX1

affinity isolated antibody

Synonym(s):

Anti- HLXB9, Anti- HOXHB9, Anti- SCRA1, Anti-HB9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_005515

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MNX1(3110)

General description

Motor neuron and pancreas homeobox 1 (MNX1) gene is located on human chromosome 7q36.3. MNX1 contains a homeodomain, which encodes transcriptional factors for both solid and hematological malignancies.

Immunogen

Synthetic peptide directed towards the N terminal region of human MNX1

Application

Anti-MNX1 produced in rabbit has been used in immunohistochemistry and western blotting.

Biochem/physiol Actions

Motor neuron and pancreas homeobox 1 (MNX1) is known to cause permanent neonatal diabetes mellitus (PNDM), which is a rare monogenic disease. In bladder cancer, MNX1 is upregulated.

Sequence

Synthetic peptide located within the following region: AAASGTGGGGGGGGASGGTSGSCSPASSEPPAAPADRLRAESPSPPRLLA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motor neuron and pancreas homeobox 1/HLXB9 promotes sustained proliferation in bladder cancer by upregulating CCNE1/2
Chen, Min, et al.
Journal of Experimental & Clinical Cancer Research, 37(1), 154-154 (2018)
Transcription factor gene MNX1 is a novel cause of permanent neonatal diabetes in a consanguineous family
Bonnefond, et al.
Diabetes and Metabolism, 39(3), 276-280 (2013)
Currarino syndrome and HPE microform associated with a 2.7-Mb deletion in 7q36. 3 excluding SHH gene
Coutton,, et al.
Molecular Syndromology, 5(1), 25-31 (2014)
The dual role of HLXB9 in leukemia
Ferguson,, et al.
Pediatric Blood & Cancer, 56(3), 349-352 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service