Skip to Content
Merck
All Photos(1)

Key Documents

SAB2105523

Sigma-Aldrich

Anti-ABCC11 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-EWWD, Anti-MRP8, Anti-WW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

154 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ABCC11(85320)

Immunogen

Synthetic peptide directed towards the middle region of human ABCC11

Biochem/physiol Actions

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, M

Sequence

Synthetic peptide located within the following region: NSKIILIDEATASIDMETDTLIQRTIREAFQGCTVLVIAHRVTTVLNCDH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine Drews-Elger et al.
Breast cancer research and treatment, 148(1), 41-59 (2014-10-02)
The mechanisms by which breast cancer (BrC) can successfully metastasize are complex and not yet fully understood. Our goal was to identify tumor-induced stromal changes that influence metastatic cell behavior, and may serve as better targets for therapy. To identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service