SAB2103055
Anti-TAF7L antibody produced in rabbit
affinity isolated antibody
Synonym(s):
Anti-FLJ23157, Anti-TAF2Q
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
51 kDa
species reactivity
pig, human
concentration
0.5 mg - 1 mg/mL
Immunogen
Synthetic peptide directed towards the middle region of human TAF7L
Biochem/physiol Actions
TAF7L gene is similar to a mouse gene that encodes a TATA box binding protein-associated factor, and shows testis-specific expression.This gene is similar to a mouse gene that encodes a TATA box binding protein-associated factor, and shows testis-specific expression.
Sequence
Synthetic peptide located within the following region: QKQIEKKEKKLHKIQNKAQRQKDLIMKVENLTLKNHFQSVLEQLELQEKQ
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service