Skip to Content
Merck
All Photos(5)

Key Documents

SAB1411127

Sigma-Aldrich

Anti-PSMD5 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA0072, MGC23145, S5B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56.2 kDa

species reactivity

rat, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSMD5(5711)

Related Categories

General description

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator base. (provided by RefSeq)

Immunogen

PSMD5 (NP_005038.1, 1 a.a. ~ 504 a.a) full-length human protein.

Sequence
MAAQALALLREVARLEAPLEELRALHSVLQAVPLNELRQQAAELRLGPLFSLLNENHREKTTLCVSILERLLQAMEPVHVARNLRVDLQRGLIHPDDSVKILTLSQIGRIVENSDAVTEILNNAELLKQIVYCIGGENLSVAKAAIKSLSRISLTQAGLEALFESNLLDDLKSVMKTNDIVRYRVYELIIEISSVSPESLNYCTTSGLVTQLLRELTGEDVLVRATCIEMVTSLAYTHHGRQYLAQEGVIDQISNIIVGADSDPFSSFYLPGFVKFFGNLAVMDSPQQICERYPIFVEKVFEMIESQDPTMIGVAVDTVGILGSNVEGKQVLQKTGTRFERLLMRIGHQSKNAPVELKIRCLDAISSLLYLPPEQQTDDLLRMTESWFSSLSRDPLELFRGISSQPFPELHCAALKVFTAIANQPWAQKLMFNSPGFVEYVVDRSVEHDKASKDAKYELVKALANSKTIAEIFGNPNYLRLRTYLSEGPYYVKPVSTTAVEGAE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service