Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406142

Sigma-Aldrich

Anti-MPP3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DLG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~66.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPP3(4356)

Related Categories

Immunogen

MPP3 (NP_001923.2, 1 a.a. ~ 585 a.a) full-length human protein.

Sequence
MPVLSEDSGLHETLALLTSQLRPDSNHKEEMGFLRDVFSEKSLSYLMKIHEKLRYYERQSPTPVLHSAVALAEDVMEELQAASVHSDERELLQLLSTPHLRAVLMVHDTVAQKNFDPVLPPLPDNIDEDFDEESVKIVRLVKNKEPLGATIRRDEHSGAVVVARIMRGGAADRSGLVHVGDELREVNGIAVLHKRPDEISQILAQSQGSITLKIIPATQEEDRLKESKVFMRALFHYNPREDRAIPCQEAGLPFQRRQVLEVVSQDDPTWWQAKRVGDTNLRAGLIPSKGFQERRLSYRRAAGTLPSPQSLRKPPYDQPCDKETCDCEGYLKGHYVAGLRRSFRLGCRERLGGSQEGKMSSGAESPELLTYEEVARYQHQPGERPRLVVLIGSLGARLHELKQKVVAENPQHFGVAVPHTTRPRKSHEKEGVEYHFVSKQAFEADLHHNKFLEHGEYKENLYGTSLEAIQAVMAKNKVCLVDVEPEALKQLRTSEFKPYIIFVKPAIQEKRKTPPMSPACEDTAAPFDEQQQEMAASAAFIDRHYGHLVDAVLVKEDLQGAYSQLKVVLEKLSKDTHWVPVSWVR

Application

Anti-MPP3 antibody produced in mouse is suitable for western blot analysis.

Biochem/physiol Actions

MPP3 (Membrane protein, palmitoylated 3, MAGUK p55 subfamily member 3) plays an important role in the maintenance of epithelial structure through cell-cell interactions. The N-terminal domain of MPP3 directly binds to the cytoplasmic domain of CADM1 (Cell adhesion molecule 1) and forms a complex, which further connects it to the PI3K (phosphoinositide 3-kinase) for its activation. The activation of PI3K is highly required to identify the actin cytoskeleton and form epithelial cell structure. A study reports that MPP3 in conjugation with TSLC1 (Tumor suppressor in lung cancer 1) shows tumor suppressive property in human lung cancer cells and any change in this interactions might lead to malignancy and tumor growth.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigefumi Murakami et al.
PloS one, 9(2), e82894-e82894 (2014-02-08)
CADM1 (Cell adhesion molecule 1), a cell adhesion molecule belonging to the immunoglobulin superfamily, is involved in cell-cell interaction and the formation and maintenance of epithelial structure. Expression of CADM1 is frequently down-regulated in various tumors derived from epithelial cells.
Hiroshi Fukuhara et al.
Oncogene, 22(40), 6160-6165 (2003-09-19)
We have previously identified the tumor suppressor in lung cancer 1 (TSLC1) gene as a novel tumor suppressor in human non-small cell lung cancer (NSCLC) by functional complementation. TSLC1 encodes a membrane glycoprotein belonging to an immunoglobulin superfamily and participates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service