Skip to Content
Merck
All Photos(1)

Key Documents

SAB1304574

Sigma-Aldrich

ANTI-TET2 (N-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

KIAA1546, Probable methylcytosine dioxygenase TET2, TET2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

223811 Da

species reactivity

human

technique(s)

western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TET2(54790)

General description

Tet methyl cytosine dioxygenase 2 (TET2), a TET oncogene family member 2/ten-eleven translocation 2, is a tumour suppressor gene which encodes a DNA demethylase enzyme. In human chromosome, the gene TET2 is localized on 4q24.

Biochem/physiol Actions

Tet methyl cytosine dioxygenase 2 (TET2) catalyses the conversion of 5-methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC) in DNA. TET2 is essential for normal myelopoiesis, epigenetic programming, embryonic stem cell maintenance and early development. Mutations in TET2 leads to myelodysplastic syndromes (MDS), myeloproliferative disorder, myeloproliferative neoplasms (MPN). Mutations also causes different leukaemia like chronic myelomonocytic leukaemia (CMML), acute myeloid leukaemias (AML), secondary AML (sAML) and myeloid malignancies. TET2 regulates mast cell differentiation and proliferation and cytokine production.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired hydroxylation of 5-methylcytosine in myeloid cancers with mutant TET2
Ko M, et al.
Nature, 468(7325), 839-839 (2010)
Leukemic IDH1 and IDH2 mutations result in a hypermethylation phenotype, disrupt TET2 function, and impair hematopoietic differentiation
Figueroa ME, et al.
Cancer Cell, 18(6), 553-567 (2010)
Mutation in TET2 in myeloid cancers
Delhommeau F, et al.
The New England Journal of Medicine, 360(22), 2289-2301 (2009)
TET2 regulates mast cell differentiation and proliferation through catalytic and non-catalytic activities
Montagner S, et al.
Cell Reports, 15(7), 1566-1579 (2016)
The role of TET2 in hematologic neoplasms
Holmfeldt L and Mullighan CG
Cancer Cell, 20(1), 1-2 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service