Skip to Content
Merck
All Photos(5)

Key Documents

HPA026846

Sigma-Aldrich

Anti-NECTIN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD111, Anti-CLPED1, Anti-ED4, Anti-HIgR, Anti-HVEC, Anti-OFC7, Anti-PRR, Anti-PRR1, Anti-PVRL1, Anti-PVRR1, Anti-SK-12, Anti-CD111 antigen, Anti-Herpes virus entry mediator C, Anti-Herpesvirus Ig-like receptor, Anti-HveC, Anti-Nectin-1, Anti-Poliovirus receptor-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

DSMYGFIGTDVVLHCSFANPLPSVKITQVTWQKSTNGSKQNVAIYNPSMGVSVLAPYRERVEFLRPSFTDGTIRLSRLELEDEGVYICEFATFPTGNRESQLNLTVMAKPTNWIEGTQAVLRAKKGQDD

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PVRL1(5818)

General description

Poliovirus receptor-related 1 (PVRL1), also known as nectin-1, is a member of immunoglobulin super-family. The gene coding for this protein is mapped to human chromosome 11q23.2. The encoded protein is characterized with ectodomain containing one V-like domain and two C2 like -domain.

Immunogen

nectin cell adhesion molecule 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Poliovirus receptor-related 1 (PVRL1) is a vital cell adhesion molecule, involved in the formation of tight junctions between epithelial cells.The encoded protein also plays a key role in secondary palate development. PVRL1 / nectin 1 is essential for proper Chlamydia trachomatis development. Mutations in PVRL1 gene results in non-syndromic cleft lip with or without cleft palate (CL/P) and ectodermal dysplasia-1. PVRL1 facilitates the entry of herpes simplex virus (HSV) 1 and 2 into human cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71778

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The V domain of herpesvirus Ig-like receptor (HIgR) contains a major functional region in herpes simplex virus-1 entry into cells and interacts physically with the viral glycoprotein D.
Cocchi F
Proceedings of the National Academy of Sciences of the USA, 95(26), 15700-15705 (1998)
Jennifer V Hall et al.
Frontiers in cellular and infection microbiology, 4, 158-158 (2014-11-22)
Interaction of Herpes Simplex Virus (HSV) glycoprotein D (gD) with the host cell surface during Chlamydia trachomatis/HSV co-infection stimulates chlamydiae to become persistent. During viral entry, gD interacts with one of 4 host co-receptors: HVEM (herpes virus entry mediator), nectin-1
Fei Wang et al.
Hypertension (Dallas, Tex. : 1979), 64(2), 369-377 (2014-05-29)
Angiotensin II (Ang II) stimulates (pro)renin receptor (PRR) expression in the renal collecting duct, triggering the local renin response in the distal nephron. Our recent study provided evidence for involvement of cyclooxygenase-2-prostaglandin E2 pathway in Ang II-dependent stimulation of PRR
Fei Wang et al.
American journal of physiology. Renal physiology, 307(1), F25-F32 (2014-04-18)
(Pro)renin receptor (PRR) is predominantly expressed in the distal nephron where it is activated by angiotensin II (ANG II), resulting in increased renin activity in the renal medulla thereby amplifying the de novo generation and action of local ANG II.
S Y Shu et al.
Genetics and molecular research : GMR, 14(2), 3400-3408 (2015-05-13)
Non-syndromic cleft of the lip and/or palate (NSCLP) is a very common birth defect; the poliovirus receptor-like 1 gene (PVRL1) has been identified as a genetic risk factor for NSCLP in patients from Norway, the Philippines, and South America. Given

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service