Skip to Content
Merck
All Photos(7)

Key Documents

HPA011912

Sigma-Aldrich

Anti-PDLIM4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LIM protein RIL, Anti-PDZ and LIM domain protein 4, Anti-Reversion-induced LIM protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HSVTLRGPSPWGFRLVGGRDFSAPLTISRVHAGSKAALAALCPGDLIQAINGESTELMTHLEAQNRIKGCHDHLTLSVSRPEGRSWPSAPDDSKAQAHRIHIDPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM4(8572)

Immunogen

PDZ and LIM domain protein 4 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

PDLIM4 (PDZ and LIM domain 4) is an actin associated protein consisting of a LIM (Lin11/Isl1/Mec3) domain and PDZ (PSD-95/Dlg/ZO-1) domain. It is generally expressed within the cytokine cluster. It is a tumor suppressor gene, which actively suppresses Src activation through interacting with Src and PTPL1 (Protein tyrosine phosphatase 1). As a result, it leads to a series of Src inactivation cycle. Therefore, silencing of the gene directly disrupts the Src inactivation cycle followed by aberrant Src activation in human cancers. It also consists of two putative transcription start points (tsp) responsible for potential binding of transcriptional factors. Mutation in the PDLIM4 is associated with radial bone mineral density (BMD) and osteoporosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71491

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie de Tayrac et al.
PloS one, 8(9), e73332-e73332 (2013-09-17)
Recent studies have highlighted the heterogeneity of gliomas and demonstrated that molecular and genetic analysis could help in their classification and in the design of treatment protocols. In a previous study we have identified a 4-gene signature highly correlated with
Olga A Guryanova et al.
The Journal of biological chemistry, 286(30), 26849-26859 (2011-06-04)
RIL (product of PDLIM4 gene) is an actin-associated protein that has previously been shown to stimulate actin bundling by interacting with actin-cross-linking protein α-actinin-1 and increasing its affinity to filamentous actin. Here, we report that the alternatively spliced isoform of
Fumihiro Omasu et al.
Journal of human genetics, 48(7), 342-345 (2003-08-09)
Twin and family studies had shown that genetic factors are important determinants of bone mass. Multiple genes might be involved. One candidate gene, the reversion-induced LIM gene ( RIL), is a PDZ and LIM-domain-containing protein and has been localized within
A A Bashirova et al.
Gene, 210(2), 239-245 (1998-05-09)
The ril gene encoding a LIM domain protein of an unknown function was previously identified by differential expression cloning as a candidate tumor suppressor gene in rat fibroblasts (Kiess, M., Scharm, B., Aguzzi, A., Hajnal, A., Klemenz, R., Schwarte-Waldhoff, I.
Yongjun Zhang et al.
The Journal of cell biology, 184(6), 785-792 (2009-03-25)
Aberrant Src activation plays prominent roles in cancer progression. However, how Src is activated in cancer cells is largely unknown. Genetic Src-activating mutations are rare and, therefore, are insufficient to account for Src activation commonly found in human cancers. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service