Skip to Content
Merck
All Photos(2)

Key Documents

HPA003345

Sigma-Aldrich

Anti-CHL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Close homolog of L1 antibody produced in rabbit, Anti-Neural cell adhesion molecule L1-like protein precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

CEFFASPEAVVSWQKVEEVKPLEGRRYHIYENGTLQINRTTEEDAGSYSCWVENAIGKTAVTANLDIRNATKLRVSPKNPRIPKLHMLELHCESKCDSHLKHSLKLSWSKDGEAFEINGTEDGRIIIDGANLTISNVTLEDQGIYCCS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHL1(10752)

General description

CHL1 (cell adhesion molecule L1-like) is also called as CALL (cell adhesion L1-like) and is expressed in normal tissues, brain, and a variety of human cancer cell lines and primary tumor tissues. The gene is mapped to human chromosome 3p26.3.

Immunogen

Neural cell adhesion molecule L1-like protein precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CHL1 (cell adhesion molecule L1-like) gene encodes a one-pass trans-membrane protein belonging to the L1 gene family of neural cell adhesion molecules. The protein is capable of both homotypic and heterotypic binding and may be involved in signal transduction pathways. The gene has been implicated in cancer growth and metastasis and some neurological diseases such as schizophrenia. It also functions in general cognitive activities. Deletion of one copy of this gene causes mental defects in patients with 3p- syndrome. It may serve as a potential biomarker in the early pathogenesis of two major histological types of renal cancer. Deficiency of this protein is found to promote tumor formation and it overexpression suppresses proliferation and invasion. It may be a potential therapeutic target for breast carcinogenesis and progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86472

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Hong He et al.
Biochemical and biophysical research communications, 438(2), 433-438 (2013-08-03)
Neural cell adhesion molecules (CAM) play important roles in the development and regeneration of the nervous system. The L1 family of CAMs is comprised of L1, Close Homolog of L1 (CHL1, L1CAM2), NrCAM, and Neurofascin, which are structurally related trans-membrane
Wei Li et al.
Journal of surgical oncology, 118(6), 1042-1049 (2018-10-13)
Hürthle cell carcinoma (HCC) is an unusual and relatively rare type of differentiated thyroid cancer. Currently, cytologic analysis of fine-needle aspiration biopsy is limited in distinguishing benign Hürthle cell neoplasms from malignant ones. The aim of this study was to
Vera N Senchenko et al.
PloS one, 6(3), e15612-e15612 (2011-03-17)
CHL1 gene (also known as CALL) on 3p26.3 encodes a one-pass trans-membrane cell adhesion molecule (CAM). Previously CAMs of this type, including L1, were shown to be involved in cancer growth and metastasis. We used Clontech Cancer Profiling Arrays (19
Jelena Katic et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(44), 14606-14623 (2014-10-31)
The cell adhesion molecule close homolog of L1 (CHL1) plays important functional roles in the developing and adult nervous system. In search of the binding partners that mediate the diverse and sometimes opposing functions of CHL1, the extracellular matrix-associated proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service