Skip to Content
Merck
All Photos(1)

Key Documents

EHU051701

Sigma-Aldrich

MISSION® esiRNA

targeting human BDKRB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCTTGAGCATCCCCACATTCCTGCTGCGATCCATCCAAGCCGTCCCAGATCTGAACATCACCGCCTGCATCCTGCTCCTCCCCCATGAGGCCTGGCACTTTGCAAGGATTGTGGAGTTAAATATTCTGGGTTTCCTCCTACCACTGGCTGCGATCGTCTTCTTCAACTACCACATCCTGGCCTCCCTGCGAACGCGGGAGGAGGTCAGCAGGACAAGGTGCGGGGGCCGCAAGGATAGCAAGACCACAGCGCTGATCCTCACGCTCGTGGTTGCCTTCCTGGTCTGCTGGGCCCCTTACCACTTCTTTGCCTTCCTGGAATTCTTATTCCAGGTGCAAGCAGTCCGAGGCTGCTTTTGGGAGGACTTCATTGACCTGGGCCTGCAATTGGCCAACTTCTTTGCCTTCACTAACAGCTCCCTGAATCCAGTAATTTATGTCTTTGTGGGCCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guojun Wang et al.
Journal of cellular biochemistry, 118(12), 4444-4453 (2017-05-04)
Bradykinin (BK) has been reported to be involved in the progression of diverse types of cancer. In the present study, we investigated the possible role of BK in cell proliferation, migration, invasion, and tumor growth of gastric cancer (GC). Cell
Ding-Ping Sun et al.
Cancers, 12(3) (2020-03-19)
Glioblastoma multiforme (GBM) is the most common form of brain tumor and is very aggressive. Rapid migration and invasion of glioblastoma cells are two typical features driving malignance of GBM. Bradykinin functionally prompts calcium influx via activation of bradykinin receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service