Skip to Content
Merck
All Photos(1)

Key Documents

AV48138

Sigma-Aldrich

Anti-PGAM2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC88743, Anti-PGAMM, Anti-Phosphoglycerate mutase 2 (muscle)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGAM2(5224)

General description

Phosphoglycerate mutase 2 (muscle) (PGAM2) catalyzes the the conversion of 3-phosphoglycrate to 2-phosphoglycerate during glucolysis. Genetic alterations in PGAM2 have been linked to muscle phosphoglycerate mutase deficiency and Greig cephalopolysyndactyly syndrome.
Rabbit Anti-PGAM2 antibody recognizes human, mouse, rat, zebrafish, and pig PGAM2.

Immunogen

Synthetic peptide directed towards the N terminal region of human PGAM2

Application

Rabbit Anti-PGAM2 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

PGAM2 is the interconversion of 3- and 2-phosphoglycerate with 2,3-bisphosphoglycerate as the primer of the reaction. It can also catalyze the reaction of EC 5.4.2.4 (synthase) and EC 3.1.3.13 (phosphatase), but with a reduced activity.

Sequence

Synthetic peptide located within the following region: MATHRLVMVRHGESTWNQENRFCGWFDAELSEKGTEEAKRGAKAIKDAKM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Wagner et al.
Genomics, 8(3), 487-491 (1990-11-01)
Greig cephalopolysyndactyly syndrome (GCPS) is an autosomal dominant disorder that has been mapped to 7p13. We have investigated two patients with GCPS and a cytogenetically visible microdeletion of the short arm of chromosome 7 with gene probes that have been

Articles

We presents an article about the Warburg effect, and how it is the enhanced conversion of glucose to lactate observed in tumor cells, even in the presence of normal levels of oxygen. Otto Heinrich Warburg demonstrated in 1924 that cancer cells show an increased dependence on glycolysis to meet their energy needs, regardless of whether they were well-oxygenated or not.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service