Skip to Content
Merck
All Photos(1)

Key Documents

AV34182

Sigma-Aldrich

Anti-CSRP3 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cysteine and glycine-rich protein 3 (cardiac LIM protein)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

rabbit, human, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSRP3(8048)

General description

CSRP3 is a LIM domain protein that may modulate cell growth and differentiation. Studies in rat wheel-lock (WL) models for depressed ambulatory activity have revealed low Csrp3 protein levels. Moreover, CSRP3 mutations have been linked to hypertrophic cardiomyopathy.
Rabbit Anti-CSRP3 recognizes human, canine, rat, mouse, bovine, and pig CSRP3.

Immunogen

Synthetic peptide directed towards the C terminal region of human CSRP3

Application

Rabbit Anti-CSRP3 can be used for western blot applications at 0.125 μg/ml.

Biochem/physiol Actions

This CSRP3 gene encodes a member of the CSRP family of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this protein is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Mutations in this gene are thought to cause heritable forms of hypertrophic cardiomyopathy (HCM) and dilated cardiomyopathy (DCM) in humans.

Sequence

Synthetic peptide located within the following region: FRCAICGKSLESTNVTDKDGELYCKVCYAKNFGPTGIGFGGLTQQVEKKE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Roberts et al.
Journal of applied physiology (Bethesda, Md. : 1985), 112(8), 1291-1299 (2012-01-28)
The wheel-lock (WL) model for depressed ambulatory activity in rats has shown metabolic maladies ensuing within 53-173 h after WL begins. We sought to determine if WL beginning after 21-23 days of voluntary running in growing female Wistar rats affected
Christian Geier et al.
Human molecular genetics, 17(18), 2753-2765 (2008-05-29)
Hypertrophic cardiomyopathy (HCM) is a frequent genetic cardiac disease and the most common cause of sudden cardiac death in young individuals. Most of the currently known HCM disease genes encode sarcomeric proteins. Previous studies have shown an association between CSRP3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service