Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
recombinant
expressed in baculovirus infected Sf21 cells
Quality Level
description
protein target CLK3
purified by
(Ni2+/NTA-agarose)
technique(s)
activity assay: suitable
UniProt accession no.
Gene Information
human ... CLK3(1198)
General description
FUNCTION: SwissProt: P49761 # Phosphorylates serine- and arginine-rich (SR) proteins of the spliceosomal complex. May be a constituent of a network of regulatory mechanisms that enable SR proteins to control RNA splicing. Phosphorylates serines, threonines and tyrosines.
SIZE: 638 amino acids; 73515 Da
SUBCELLULAR LOCATION: Nucleus. Cytoplasm (By similarity). Acrosome (By similarity).
PTM: Autophosphorylates on all three types of residues (By similarity).
SIMILARITY: SwissProt: P49761 ## Belongs to the protein kinase superfamily. CMGC Ser/Thr protein kinase family. Lammer subfamily. & Contains 1 protein kinase domain.
SIZE: 638 amino acids; 73515 Da
SUBCELLULAR LOCATION: Nucleus. Cytoplasm (By similarity). Acrosome (By similarity).
PTM: Autophosphorylates on all three types of residues (By similarity).
SIMILARITY: SwissProt: P49761 ## Belongs to the protein kinase superfamily. CMGC Ser/Thr protein kinase family. Lammer subfamily. & Contains 1 protein kinase domain.
Quality
routinely evaluated by phosphorylation of 500μM PKC Substrate Peptide 2 (Catalog # 12-411)
Physical properties
63kDa
Storage and Stability
6 months at -70°C from date of shipment
Analysis Note
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service