GF85362891
Zirconium
foil, light tested, 50x50mm, thickness 0.007mm, 99.2%
Synonym(s):
Zirconium, ZR000140
Select a Size
Select a Size
About This Item
Assay
99.2%
form
foil
manufacturer/tradename
Goodfellow 853-628-91
resistivity
40 μΩ-cm, 20°C
size × thickness
50 x 50 mm × 0.007 mm
bp
4377 °C (lit.)
mp
1852 °C (lit.)
density
6.5 g/mL at 25 °C (lit.)
SMILES string
[Zr]
InChI
1S/Zr
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service