GF82636162
Copper
tube, 100mm, outside diameter 25.4mm, inside diameter 18.9mm, wall thickness 3.25mm, as drawn, 99.9%
Synonym(s):
Copper, CU007700
About This Item
Assay
99.9%
form
tubes
manufacturer/tradename
Goodfellow 826-361-62
resistivity
1.673 μΩ-cm, 20°C
L × wall thickness
100 mm × 3.25 mm
O.D. × I.D.
25.4 mm × 18.9 mm
bp
2567 °C (lit.)
mp
1083.4 °C (lit.)
density
8.94 g/mL at 25 °C (lit.)
SMILES string
[Cu]
InChI
1S/Cu
InChI key
RYGMFSIKBFXOCR-UHFFFAOYSA-N
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service