GF68076490
Stainless Steel - 17-7PH alloy, FeCr17Ni7
foil, 150 x 150mm, 0.075mm thickness, annealed - condition a, precipitation hardenable, light tight (lt)
Synonym(s):
FE280300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
description
light tight (lt)
form
annealed (condition a: precipitation hardenable
quarter hard)
manufacturer/tradename
Goodfellow 680-764-90
size × thickness
150 mm × 150 mm × 0.075 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service