GF62806305
Silver
foil, 4mm disks, thickness 0.1mm, annealed, 99.95+%
Synonym(s):
Silver, AG000351
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Assay
99.95%
form
foil
manufacturer/tradename
Goodfellow 628-063-05
resistivity
1.59 μΩ-cm, 20°C
diam. × thickness
4 mm × 0.1 mm
bp
2212 °C (lit.)
mp
960 °C (lit.)
density
10.49 g/cm3 (lit.)
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
IX. Note on Silver Foil in Surgery.
J S Lewis
Annals of surgery, 50(4), 793-796 (1909-10-01)
Clinical evaluation of C-Film, a vaginal contraceptive.
O Frankman et al.
The Journal of international medical research, 3(4), 292-296 (1975-01-01)
H H Wiebe et al.
Plant physiology, 59(2), 256-258 (1977-02-01)
Water potential was monitored at nine locations along single maize (Zea mays L.) leaf blades with aluminum block in situ thermocouple hygrometers. Water potential showed a continuous decrease toward the tip, with a 2- to 4-bar difference between leaf base
A Rupérez et al.
Talanta, 44(2), 213-220 (1997-02-01)
Surface-enhanced Raman scattering (SERS) spectrometry of the diuretic drug triamterene is discussed. The SERS-active substrate used is a silver foil etched with nitric acid. The influence of solvent and sample doping method on sensivity, intercept and shape of the calibration
M Dubiel et al.
Journal of synchrotron radiation, 8(Pt 2), 539-541 (2001-08-22)
The thermal expansion behaviour of silver fcc has been described by an anharmonic Einstein model using EXAFS data in the temperature range between 10 and 300 K. The linear expansion coefficient of a bulk silver foil agrees well with X-ray
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service