Skip to Content
Merck
All Photos(1)

Key Documents

DWK218016957

DURAN® laboratory bottles, with caps

new

bottle capacity 3500mL, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121818

material

borosilicate glass 3.3 bottle
clear
polypropylene cap

Joint

joint threaded neck

sterility

non-sterile

feature

autoclavable
graduated
with cap
pouring ring

packaging

pack of 1 ea

manufacturer/tradename

DWK Life Sciences DURAN® 218016957

bottle capacity

3500mL

bottle diam. × H

160 mm × 300 mm

fitting

GL 45 thread

Looking for similar products? Visit Product Comparison Guide

General description

DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN® Original GL bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140 °C. All components are fully autoclavable at 121 or 134 °C.

Application

DURAN® graduated laboratory bottles are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.

Features and Benefits

  • Complies with the standards in ISO 4796-1:2016 Laboratory glassware -- Bottles -- Part 1: Screw-neck bottles (does not apply to 10 ml bottle)
  • Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • Retrace Code, and certificate for manufacturing lot traceability
  • Fired-on, highly durable white print
  • Suitable for long-term storage
  • Easy-to-read scale with graduation marks to estimate the content

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service