DWK213524104
Duran® Funnel Ribbed
Synonym(s):
213524104
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 213524104
funnel diam.
80 mm
Looking for similar products? Visit Product Comparison Guide
Application
Decanting and filtration of liquids, The spillage of substances is avoided, Filtering, Boro 3.3
Features and Benefits
Due to its shape, it is suitable for liquid-solid filtration using round filter paper; Fired-on,
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service