Bioprocessing Bioreactors
The Mobius® family of single-use bioreactors is a scalable portfolio that includes benchtop (3 L), pilot scale (50 L and 200 L), and clinical and commercial scale (1,000 L and 2,000 L) stirred tank bioreactors, enabling optimized cell culture from early process development through commercial production. Mobius® bioreactors accommodate your fed-batch or perfusion applications with flexible configuration of software, hardware and single-use assemblies, ensuring that operational flexibility at small scale can be translated to full-scale production.
Key advantages include:
- Simple installation of Flexware® assemblies by a single operator
- 5:1 turndown ratio at process scale
- Works as a stand-alone system or integrated as part of your facility’s automation platform
- Demonstrated scalability of key engineering parameters gives you confidence to scale up your process
- Lynx® connectors assure critical sterile connections
- Mobius® SensorReady Technology for bioreactor monitoring and control
Webinar: Scalability and Robustness of the Mobius® Bioreactor Platform
Application Note: Scalability of the Mobius® Single-use Bioreactors
Specification Sheet: Mobius® Bioreactors: A Scalable, Ergonomic System with the Flexibility to Fit Your Process
Poster: Utility of the Mobius® 3 L Single-use Bioreactor for Upstream Process Development Applications