SIAL0166
Sigma® cell culture dish
diam. × H 60 mm × 15 mm, tissue-culture treated, sterile, pack of 25 × 20 ea
About This Item
Kod UNSPSC:
41121812
NACRES:
NB.22
Pomoc techniczna
Pozwól nam pomócPomoc techniczna
Pozwól nam pomócsterylność
sterile
opakowanie
pack of 25 × 20 ea
śr. × wys.
60 mm × 15 mm
powierzchnia
21 cm2
typ wiązania
Tissue Culture (TC)-treated surface
Opis ogólny
- Manufactured from optically-clear virgin polystyrene
- Treated for optimal cell attachment
- Sterilized by gamma radiation and certified nonpyrogenic
- Have stacking beads to aid in handling
- Lid supplied with vents to provide consistent gas exchange
Informacje prawne
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under Sigma® cell culture dish
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?