Skip to Content
Merck

63301-001

Sigma-Aldrich

PCRboost®

Trial Kit, for improved PCR performance

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

usage

3 reactions

storage temp.

room temp

General description

PCRboost® is a unique reagent that enhances end-point and reverse transcription-PCR performance by improving sensitivity and specificity during amplification of genomic DNA or RNA templates. To use, simply replace the water in your PCR with PCRboost®. That′s all. It is designed to enhance the amplification of DNA without the need for time consuming optimizations. PCRboost® successfully amplifies thermally degraded DNA resulting in a visible amplicon that can be used for downstream applications such as cloning and sequencing.

Don′t see what you are looking for? Custom stabilization services for molecular diagnostic assay and reagent manufacturers are available through Sigma′s partner, Biomatrica. PCRstable® enables the preservation of PCR-based assay components in any format (liquid or dry).

Packaging

1 bottle

Physical form

150 μL of PCRboost® in 1.5 mL screw-top conical tube

Legal Information

PCRboost is a registered trademark of Biomatrica
PCRstable is a registered trademark of Biomatrica


Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library