Skip to Content
Merck
Iron(II) chloride anhydrous ≥99.99% trace metals basis

Iron(II) chloride anhydrous

Iron(II) chloride anhydrous ≥99.99% trace metals basis
Synonym(s):
Iron(II) chloride, Ferrous chloride, Iron dichloride
Linear Formula:
FeCl2
CAS Number:
Molecular Weight:
126.75

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing