Skip to Content
Merck
All Photos(1)

Key Documents

V900915

Protein Marker

Vetec, reagent grade, low range, mol wt 12,000-80,000 Da

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

reagent grade

product line

Vetec

form

solution

usage

sufficient for 10-20 large gels (16 × 18 cm)
sufficient for 20-30 mini-gels (10 × 10 cm)
sufficient for 50-100 PhastGels

mol wt

12,000-80,000 Da

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For use with Laemmli SDS-PAGE, the PhastSystem (Cytiva) and Schagger and von Jagow SDS-PAGE (Tricine). The markers are ready-to-use solutions formulated to yield a distribution of well-defined bands of approximately equal intensity after electrophoresis and Coomassie Blue staining.
Will yield 6 protein bands with Coomassie Blue staining (V900852).

Other Notes

6 polypeptides bands as 12 kDa, 20 kDa, 30 kDa, 40 kDa, 60 kDa, 80 kDa.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabina Honisch et al.
Oncotarget, 6(12), 10309-10319 (2015-04-15)
Chorein encoded by VPS13A (vacuolar protein sorting-associated protein 13A) is defective in chorea-acanthocytosis. Chorein fosters neuronal cell survival, cortical actin polymerization and cell stiffness. In view of its anti-apoptotic effect in neurons, we explored whether chorein is expressed in cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service