Skip to Content
Merck
All Photos(3)

Key Documents

V900870

Sigma-Aldrich

Iodonitrotetrazolium chloride

Vetec, reagent grade, ≥98%

Synonym(s):

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, p-Iodonitrotetrazolium Violet, INT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClIN5O2
CAS Number:
Molecular Weight:
505.70
Beilstein:
4093224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

mp

240 °C (dec.) (lit.)

solubility

H2O: 4 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].[O-][N+](=O)c1ccc(cc1)-[n+]2nc(nn2-c3ccc(I)cc3)-c4ccccc4

InChI

1S/C19H13IN5O2.ClH/c20-15-6-8-16(9-7-15)23-21-19(14-4-2-1-3-5-14)22-24(23)17-10-12-18(13-11-17)25(26)27;/h1-13H;1H/q+1;/p-1

InChI key

JORABGDXCIBAFL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Electron acceptor for the colorimetric assay of various dehydrogenases

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinghui Zhang et al.
Cancer investigation, 33(5), 165-171 (2015-04-02)
This study demonstrated that miR-21 and miR-95 expression were significantly higher in the ALDH1(+)CD133(+)subpopulation than in the ALDH1(-)CD133(-) subpopulation of lung cancer cells. Combined delivery of anti-miR-21 and anti-miR-95 by calcium phosphate nanoparticles significantly inhibited tumor growth in a xenograft
Hsiu-Hua Wang et al.
Archives of toxicology, 88(8), 1549-1559 (2014-02-27)
We previously reported that the sustained exposure of human urothelial cells (HUCs) to low-dose sodium arsenite induces changes in the gene expression profile and neoplastic transformation. In this study, we used the HumanMethylation27 BeadChip to analyze genome-wide methylation profiles and
Jiahui Zhou et al.
Experimental biology and medicine (Maywood, N.J.), 240(7), 867-875 (2015-01-13)
This study investigated the role of miR-143 in the chemoresistance of osteosarcoma tumor cells and the associated mechanisms. Real-time PCR was used to measure miR-143 levels. Western blot was used to detect protein expression. Cell proliferation was analyzed by MTT
Iawen Hsu et al.
Oncotarget, 5(17), 7917-7935 (2014-10-04)
Clinical reports show males have a higher bladder cancer (BCa) incidence than females. The sexual difference of BCa occurrence suggests that estrogen and its receptors may affect BCa development. Estrogen receptor alpha (ERα) is the classic receptor to convey estrogen
Teng Chang et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 103(3), 539-547 (2014-06-18)
Hemocompatibility of genistein-modified poly(ethersulfone)/poly(vinylpyrrolidone) (PES/PVP) hemodialysis (HD) membranes has been investigated in vitro with emphasis on evaluation of cell viability, antioxidant, anti-inflammatory, and antiplatelet adhesion properties. Genistein modified PES/PVP membranes reveal significant reduction of the reactive oxygen species and also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service