Skip to Content
Merck
All Photos(3)

Key Documents

V900715

Sigma-Aldrich

Quinine monohydrochloride dihydrate

Vetec, reagent grade, 88%

Synonym(s):

Quinine hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · HCl · 2H2O
CAS Number:
Molecular Weight:
396.91
Beilstein:
6112655
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

88%

mp

115-116 °C (dec.) (lit.)

SMILES string

Cl[H].[H]O[H].[H]O[H].[H][C@@]12CCN(C[C@@H]1C=C)[C@@]([H])(C2)[C@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H24N2O2.ClH.2H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;2*1H2/t13-,14-,19-,20+;;;/m0.../s1

InChI key

MPQKYZPYCSTMEI-FLZPLBAKSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service