Skip to Content
Merck
All Photos(3)

Key Documents

V900481

Sigma-Aldrich

Cesium chloride

Vetec, reagent grade, ≥99%

Synonym(s):

CsCl, cesium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
reagent grade

grade

reagent grade

product line

Vetec

Assay

≥99%

mp

645 °C (lit.)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Jurkiewicz et al.
Biochimica et biophysica acta, 1818(3), 609-616 (2011-12-14)
Effects of alkali metal chlorides on the properties of mixed negatively charged lipid bilayers are experimentally measured and numerically simulated. Addition of 20mol% of negatively charged phosphatidylserine to zwitterionic phosphatidylcholine strengthens adsorption of monovalent cations revealing their specificity, in the
Therese Abrahamsson et al.
Neuron, 73(6), 1159-1172 (2012-03-27)
Interneurons are critical for neuronal circuit function, but how their dendritic morphologies and membrane properties influence information flow within neuronal circuits is largely unknown. We studied the spatiotemporal profile of synaptic integration and short-term plasticity in dendrites of mature cerebellar
Takafumi Yamamoto et al.
Inorganic chemistry, 50(22), 11787-11794 (2011-10-25)
The rock salt (B1) structure of binary oxides or chalcogenides transforms to the CsCl (B2) structure under high pressure, with critical pressures P(s) depending on the cation to anion size ratio (R(c)/R(a)). We investigated structural changes of A(2)MO(3) (A =
Shengyuan Ding et al.
Journal of neurophysiology, 106(6), 3019-3034 (2011-09-02)
GABA projection neurons (GABA neurons) in the substantia nigra pars reticulata (SNr) and dopamine projection neurons (DA neurons) in substantia nigra pars compacta (SNc) have strikingly different firing properties. SNc DA neurons fire low-frequency, long-duration spikes, whereas SNr GABA neurons
Raphaele Germi et al.
The Journal of general virology, 93(Pt 8), 1769-1773 (2012-05-18)
Epstein-Barr virus (EBV), a gammaherpesvirus, infects >90 % of the world's population. Primary infection by EBV can lead to infectious mononucleosis, and EBV persistence is associated with several malignancies. Despite its importance for human health, little structural information is available on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service