Skip to Content
Merck
All Photos(1)

Key Documents

V800416

Sigma-Aldrich

tert-Butanol

AR

Synonym(s):

2-Methyl-2-propanol, tert-Butyl alcohol, Trimethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
Beilstein:
906698
EC Number:
MDL number:
UNSPSC Code:
12191601
PubChem Substance ID:
grade:
AR
bp:
83 °C (lit.)
vapor pressure:
31 mmHg ( 20 °C)
44 mmHg ( 26 °C)

grade

AR

vapor density

2.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)
44 mmHg ( 26 °C)

product line

Vetec

form

liquid

autoignition temp.

896 °F

expl. lim.

8 %

dilution

(for analytical testing)

refractive index

n20/D 1.387 (lit.)

pH

7 (20 °C)

bp

83 °C (lit.)

mp

23-26 °C (lit.)

density

0.775 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)O

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3

InChI key

DKGAVHZHDRPRBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnau Bassegoda et al.
Applied microbiology and biotechnology, 97(19), 8559-8568 (2013-01-22)
Rhodococci are highly adaptable bacteria, capable to degrade or transform a large number of organic compounds, including recalcitrant or toxic products. However, little information is available on the lipases of the genus Rhodococcus, except for LipR, the first lipase isolated
Ee Ling Yong et al.
Water research, 46(6), 1990-1998 (2012-02-14)
Ozonation is widely employed in water treatment to purify water. The R(ct) concept, which is defined as the ratio of OH exposure to ozone exposure, has been commonly used to quantify the OH concentration generating from ozone decomposition and model
Jaime Salazar et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 81(1), 82-90 (2012-01-12)
Nanosizing is a non-specific approach to improve the oral bioavailability of poorly soluble drugs. The decreased particle size of these compounds results in an increase in surface area. The outcome is an increased rate of dissolution, which can lead to
Tesfamichael H Kebrom et al.
Plant, cell & environment, 38(8), 1471-1478 (2014-12-17)
Shoot branches or tillers develop from axillary buds. The dormancy versus outgrowth fates of buds depends on genetic, environmental and hormonal signals. Defoliation inhibits bud outgrowth indicating the role of leaf-derived metabolic factors such as sucrose in bud outgrowth. In
Jannis Wenk et al.
Environmental science & technology, 49(14), 8541-8549 (2015-06-20)
Dissolved organic matter (DOM) is both a promoter and an inhibitor of triplet-induced organic contaminant oxidation. This dual role was systematically investigated through photochemical experiments with three types of DOM of terrestrial and aquatic origins that were preoxidized to varying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service