Skip to Content
Merck
All Photos(1)

Key Documents

V800399

Sigma-Aldrich

Sodium phosphate monobasic dihydrate

LR, ≥98%

Synonym(s):

Sodium dihydrogen phosphate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
LR
form:
solid

grade

LR

product line

Vetec

Assay

≥98%

form

solid

pH

4.0-4.5 (25 °C, 50 g/L)

SMILES string

O.O.[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1

InChI key

VBJGJHBYWREJQD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Escuder-Gilabert et al.
Journal of chromatography. A, 1363, 331-337 (2014-07-22)
The design of experiments (DOE) is a good option for rationally limiting the number of experiments required to achieve the enantioresolution (Rs) of a chiral compound in capillary electrophoresis. In some cases, the modeled Rs after DOE analysis can be
Wenhu Zhou et al.
Analytical chemistry, 87(7), 4001-4007 (2015-03-11)
RNA-cleaving DNAzymes provide a unique platform for developing biosensors. However, a majority of the work has been performed in clean buffer solutions, while the activity of some important DNAzymes in biological sample matrices is still under debate. Two RNA-cleaving DNAzymes
Jan Muendges et al.
Biotechnology progress, 31(4), 925-936 (2015-04-11)
This article presents results of continuous multistage aqueous two-phase extraction of an immunoglobulin G1 from cell supernatant in a mixer-settler unit. An aqueous two-phase system consisting of polyethylene glycol 2000, phosphate salt, and water was applied without and with sodium
Soon Sik Kwon et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 92, 146-154 (2015-03-11)
We investigated the physicochemical properties of pH-sensitive hydroxyethyl cellulose (HEC)/hyaluronic acid (HA) complex hydrogels containing isoliquiritigenin (ILTG), and discussed potential applications as transdermal delivery systems for the treatment of skin lesions caused by pH imbalance. HA has skin compatibility and
Ceren Erdem et al.
Artificial cells, nanomedicine, and biotechnology, 42(4), 237-244 (2013-06-26)
In the present work, we designed an amperometric glucose biosensor based on nickel oxide nanoparticles (NiONPs)-modified carbon paste electrode. The biosensor was prepared by incorporation of glucose oxidase and NiONPs into a carbon paste matrix. It showed good analytical performances

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service