Skip to Content
Merck
All Photos(1)

Key Documents

V800388

Sigma-Aldrich

Sodium disulfite

AR, 58.5% SO2 basis

Synonym(s):

Sodium metabisulfite, Sodium disulfite, Sodium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
powder

grade

AR

product line

Vetec

form

powder

concentration

58.5% SO2

pH

4.5 (20 °C, 50 g/L)

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

HRZFUMHJMZEROT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolett Czipa et al.
Journal of food science and technology, 56(3), 1245-1255 (2019-04-09)
This study investigated the effect of different treatments (centrifugation and filtration; heating; adulteration with sugar syrups, and storage) and collection variables (year and region of the country) on the physicochemical properties of 44 Hungarian acacia honeys. The characteristics measured were
B B Gengenbach et al.
Protein expression and purification, 151, 46-55 (2018-06-13)
Cardiovascular diseases are a prevalent cause of morbidity and mortality especially in industrialized countries. The human phosphatase and actin regulator 1 (PHACTR1) may be involved in such diseases, but its precise regulatory function remains unclear due to the large number
J Y Zhu et al.
Bioresource technology, 102(19), 8921-8929 (2011-08-10)
Native aspen (Populus tremuloides) was pretreated using sulfuric acid and sodium bisulfite (SPORL) and dilute sulfuric acid alone (DA). Simultaneous enzymatic saccharification and fermentation (SSF) was conducted at 18% solids using commercial enzymes with cellulase loadings ranging from 6 to
Fiona M F Lun et al.
Clinical chemistry, 59(11), 1583-1594 (2013-07-17)
Epigenetic mechanisms play an important role in prenatal development, but fetal tissues are not readily accessible. Fetal DNA molecules are present in maternal plasma and can be analyzed noninvasively. We applied genomewide bisulfite sequencing via 2 approaches to analyze the
Lihong Fan et al.
International journal of biological macromolecules, 50(1), 31-37 (2011-10-15)
Quaternary ammonium chitosan sulfates with diverse degrees of substitution (DS) ascribed to sulfate groups between 0.52 and 1.55 were synthesized by reacting quaternary ammonium chitosan with an uncommon sulfating agent (N(SO(3)Na)(3)) that was prepared from sodium bisulfite (NaHSO(3)) through reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service