Skip to Content
Merck
All Photos(1)

Key Documents

V800315

Sigma-Aldrich

Potassium dichromate

LR, ≥99.5%

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

3.6 (100 g/L)

mp

398 °C (lit.)

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter A Siver et al.
American journal of botany, 102(6), 921-941 (2015-06-24)
Heterokont algae of the class Synurophyceae, characterized by distinctive siliceous scales that cover the surface of the cell, are ecologically important in inland waters, yet their evolutionary history remains enigmatic. We explore phylogenetic relationships within this group of algae relative
Eduardo Molina-Jijón et al.
Toxicology, 291(1-3), 93-101 (2011-11-26)
Deferoxamine (DFO) is a recognized iron chelator which has been shown to exert nephroprotection in models of toxic nephropathies. In the present work the potential protective effects of DFO against Cr(VI)-induced nephrotoxicity and oxidant stress were evaluated. Rats were injected
B Duarte et al.
Ecotoxicology and environmental safety, 83, 1-7 (2012-07-13)
The in situ reduction of Cr (VI) to its less toxic form Cr (III) may be a useful detoxification mechanism for phytoremediation. Using a hydroponics mesocosmos approach, we evaluated the ability of Halimione portulacoides to reduce and uptake Cr (VI)
Athanasia G Tekerlekopoulou et al.
Water research, 47(2), 623-636 (2012-11-28)
The kinetics of hexavalent chromium bio-reduction in draw-fill suspended and attached growth reactors was examined using sugar as substrate and indigenous microorganisms from the industrial sludge of the Hellenic Aerospace Industry. Initially, experiments in suspended growth batch reactors for Cr
Bour-Jr Wang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 110(12), 775-779 (2012-01-18)
Occupational dermatitis among cement workers is a major occupational health concern. The two most important occupational hazards for cement workers are irritant and allergic cement contact dermatitis. The objective of this study was to investigate the severity of occupational cement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service