About This Item
80-100 °C
7.99 psi ( 20 °C)
Recommended Products
grade
LR
vapor density
2.5 (vs air)
vapor pressure
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)
product line
Vetec™
autoignition temp.
475 °F
expl. lim.
8 %
dilution
(for analytical testing)
refractive index
n20/D 1.363 (lit.)
bp
30-60 °C (lit.)
80-100 °C
density
0.64 g/mL at 25 °C (lit.)
InChI
1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1
InChI key
ZRJNGFJIBZKXTP-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-56.2 °F - closed cup
Flash Point(C)
-49 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service