Skip to Content
Merck
All Photos(1)

Key Documents

V800270

Supelco

N,N-Dimethylacetamide

suitable for HPLC, ≥99.7%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

grade

HPLC grade

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

product line

Vetec

Assay

≥99.7%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

technique(s)

HPLC: suitable

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inês Farinha et al.
Carbohydrate polymers, 130, 455-464 (2015-06-17)
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic
André H Gröschel et al.
Nature communications, 3, 710-710 (2012-03-20)
Hierarchical self-assembly offers elegant and energy-efficient bottom-up strategies for the structuring of complex materials. For block copolymers, the last decade witnessed great progress in diversifying the structural complexity of solution-based assemblies into multicompartment micelles. However, a general understanding of what
Fen Ran et al.
Acta biomaterialia, 7(9), 3370-3381 (2011-06-11)
An amphiphilic triblock co-polymer of poly(vinyl pyrrolidone)-b-poly(methyl methacrylate)-b-poly(vinyl pyrrolidone) (PVP-b-PMMA-b-PVP) was synthesized via reversible addition-fragmentation chain transfer (RAFT) polymerization. The block co-polymer can be directly blended with polyethersulfone (PES) using dimethylacetamide (DMAC) as the solvent to prepare flat sheet and
S Sood et al.
Theriogenology, 78(7), 1557-1569 (2012-09-18)
Three experiments were conducted to optimize the protocol for cryopreservation of emu sperm. Ejaculates were collected from trained male emus then diluted 1:1 and pooled before allocation to treatments and measured for sperm viability, motility, egg membrane penetration ability, membrane
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service