Skip to Content
Merck
All Photos(1)

Key Documents

V800238

Sigma-Aldrich

Lead oxide, red

LR, ≥85%

Synonym(s):

Lead(II,IV) oxide, Minium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb3O4
CAS Number:
Molecular Weight:
685.60
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
powder

grade

LR

vapor pressure

10 mmHg ( 0 °C)

product line

Vetec

Assay

≥85%

form

powder

SMILES string

O1[PbH2]O[Pb]12O[PbH2]O2

InChI

1S/4O.3Pb

InChI key

XMFOQHDPRMAJNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Ox. Sol. 2 - Repr. 1A - STOT RE 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azarcón and empacho.
A Ackerman et al.
Pediatrics, 73(1), 114-115 (1984-01-01)
Lead oxide emissions--a case history.
J Delaine
The Annals of occupational hygiene, 30(2), 257-261 (1986-01-01)
L Yáñez et al.
Journal of ethnopharmacology, 41(1-2), 91-97 (1994-01-01)
Azarcon, a lead tetroxide salt, is used among Mexican and Mexican-American populations for the treatment of digestive illness. Chemical analysis of the azarcon sample used in this study showed it to be 96% lead, 1% calcium, 1% other minor metals
R D Baer et al.
Journal of ethnopharmacology, 24(1), 31-39 (1988-09-01)
This article discusses the availability of three lead-based salts which are being used as folk remedies in Mexico. Distribution systems and geographic availability were determined, and purchased samples were found to be very high in lead content. The findings suggest
Leads from the MMWR. Lead poisoning from Mexican folk remedies--California.
JAMA, 250(23), 3149-3149 (1983-12-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service