Skip to Content
Merck
All Photos(1)

Key Documents

V800237

Sigma-Aldrich

Lead(II) nitrate

LR, ≥99%

Synonym(s):

Lead dinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
LR
form:
solid

grade

LR

vapor pressure

<0.1 hPa ( 20 °C)

product line

Vetec

Assay

≥99%

form

solid

pH

3-4 (20 °C, 50 g/L)

mp

470 °C (dec.) (lit.)

SMILES string

[PbH2++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2

InChI key

RLJMLMKIBZAXJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Several new polymeric complexes of lead with dialkylamino derivatives of squaric acid were produced to study their coordination modes as possible prototypes for studies of lanthanide complexes.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trivedi Mrugesh et al.
Acta poloniae pharmaceutica, 68(5), 653-656 (2011-09-21)
The present study was an attempt to investigate toxic effects of lead on red blood corpuscle (RBC) under in vitro conditions. Venous blood samples from adult, healthy, well-nourished male donors falling in age group of 25-30 years were collected in
Merylin Zizza et al.
Aquatic toxicology (Amsterdam, Netherlands), 126, 231-241 (2012-12-19)
The hindering effects of metals and in particular lead (Pb) are representing a growing threat to aquatic organisms such as fish. This observation derives from toxic concentrations of Pb accounting for altered neurophysiological activities of some interesting teleost models like
Mostafa Lamhamdi et al.
Comptes rendus biologies, 334(2), 118-126 (2011-02-22)
Lead (Pb) is an environmental pollutant extremely toxic to plants and other living organisms including humans. To assess Pb phytotoxicity, experiments focusing on germination of wheat seeds were germinated in a solution containing Pb (NO(3))(2) (0.05; 0.1; 0.5; 1g/L) during
Hans J Jakobsen et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 211(2), 195-206 (2011-06-28)
The acquisition and different appearances observed for wide bandwidth solid-state MAS NMR spectra of low-γ nuclei, using (14)N as an illustrative nucleus and employing two different commercial spectrometers (Varian, 14.1T and Bruker, 19.6T), have been compared/evaluated and optimized from an
Xuemei Li et al.
Journal of hazardous materials, 213-214, 55-61 (2012-02-24)
An endophytic fungus was tested in rice (Oryza sativa L.) exposed to four levels of lead (Pb) stress (0, 50, 100 and 200 μM) to assess effects on plant growth, photosynthesis and antioxidant enzyme activity. Under Pb stress conditions, endophyte-infected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service