Skip to Content
Merck
All Photos(1)

Key Documents

V800219

Sigma-Aldrich

Isopentyl acetate

LR, ≥95%

Synonym(s):

Acetic acid 3-methylbutyl ester, Isoamyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744750
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
LR
bp:
142 °C/756 mmHg (lit.)
vapor pressure:
5 mmHg ( 25 °C)

grade

LR

vapor density

4.5 (vs air)

vapor pressure

5 mmHg ( 25 °C)

product line

Vetec

Assay

≥95%

form

liquid

autoignition temp.

680 °F

expl. lim.

7.5 %

dilution

(for analytical testing)

refractive index

n20/D 1.4 (lit.)

bp

142 °C/756 mmHg (lit.)

mp

−78 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCOC(C)=O

InChI

1S/C7H14O2/c1-6(2)4-5-9-7(3)8/h6H,4-5H2,1-3H3

InChI key

MLFHJEHSLIIPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gul Ozyilmaz et al.
Food chemistry, 135(4), 2326-2332 (2012-09-18)
Candida rugosa lipase (CRL) and porcine pancreatic lipase (PPL) were immobilised by entrapping and also by covalent binding for use in synthesis of isoamyl acetate (IAAc), which has a typical banana flavour. Lipase entrapment was carried out by dripping sodium
Tatjana Schreder et al.
Rhinology, 46(3), 175-183 (2008-10-16)
The aim of this study was to compare the olfactory performance of anorectic patients and healthy controls with regard to the state of satiety. Using the Sniffin' Sticks, sensitivity to a nonfood odor (n-butanol) and to a food-related odor (isoamyl
J Albrecht et al.
Rhinology, 47(2), 160-165 (2009-07-15)
The primary aim of this study was to investigate whether olfactory detection thresholds are dependent on different states of satiety. Using the threshold test of the Sniffin' Sticks test battery (single-staircase, three alternative forced choice procedure), sensitivity to a non-food
Gianfranco Anfora et al.
Behavioural brain research, 206(2), 236-239 (2009-09-22)
Recent evidence suggests that asymmetry between the left and right sides of the brain is not limited to vertebrates but extends to invertebrates as well. We compared olfactory lateralization in two species of Hymenoptera Apoidea, the honeybee (Apis mellifera), a
A M Kleemann et al.
Physiology & behavior, 97(3-4), 401-405 (2009-03-24)
The human ability to localize odorants has been examined in a number of studies, but the findings are contradictory. In the present study we investigated the human sensitivity and ability to localize hydrogen sulphide (H(2)S), which in low concentrations stimulates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service