Skip to Content
Merck
All Photos(1)

Key Documents

V800184

Sigma-Aldrich

Ferroin indicator solution

2 % (w/v)

Synonym(s):

1,10-Phenanthroline iron(II) sulfate complex, o-Phenanthroline ferrous sulfate complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Fe(C12H8N2)3]SO4
CAS Number:
Molecular Weight:
692.52
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

product line

Vetec

Quality Level

concentration

2 % (w/v)

SMILES string

[Fe++].[O-]S([O-])(=O)=O.c1cnc2c(c1)ccc3cccnc23.c4cnc5c(c4)ccc6cccnc56.c7cnc8c(c7)ccc9cccnc89

InChI

1S/3C12H8N2.Fe.H2O4S/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;1-5(2,3)4/h3*1-8H;;(H2,1,2,3,4)/q;;;+2;/p-2

InChI key

CIWXFRVOSDNDJZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Chen et al.
Guang pu xue yu guang pu fen xi = Guang pu, 25(3), 439-443 (2005-07-15)
A micro biochemical analytical system based on a micro fiber spectrometer is introduced. Experiment was carried out to calibrate and test the analysis system. In the experiment, the absorption spectra of Fe2+ -ferroin solution bodies with different concentrations were obtained.
Melanie M Britton et al.
The journal of physical chemistry. A, 109(37), 8306-8313 (2006-07-13)
The formation of stationary concentration patterns in a packed-bed reactor (PBR), using a manganese-catalyzed Belousov-Zhabotinsky (BZ) reaction in a mixed sulfuric-phosphoric acid medium, was studied using magnetic resonance imaging (MRI). The PBR was composed of a column filled with glass
E Plantin-Carrenard et al.
Annales de biologie clinique, 62(4), 456-461 (2004-08-07)
Iodine is an essential element for thyroid hormone synthesis. Iodine disorders induced biological and/to clinical expression of thyroid dysfunction. Inappropriate iodine intake (by default or by excess) is worrying in terms of public health in France regarding the iodine deficiency
S L Panwar et al.
Yeast (Chichester, England), 18(12), 1117-1129 (2001-09-06)
We report the isolation of a novel C. albicans gene designated CaALK8, by its ability to complement drug hypersensitivity of a pdr5 (ABC: ATP-binding cassette drug extrusion pump) null mutant of S. cerevisiae (JG436). CaALK8 in JG436 conferred resistance to
Chad T Hamik et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 65(4 Pt 2A), 046224-046224 (2002-05-15)
We report experimental results on the dynamics of excitation waves in a modified Belousov-Zhabotinsky reaction. The waves in this system obey nonmonotonic dispersion relations. This anomaly induces the stacking of excitation fronts into patterns with stable interpulse distances. The stacking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service