Skip to Content
Merck
All Photos(1)

Key Documents

V800159

Sigma-Aldrich

Diethylamine

LR, ≥99%

Synonym(s):

N,N-Diethylamine, N-ethyl-ethanamine, DEA, Diethamine, Diethyl amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
Beilstein:
605268
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
bp:
55 °C (lit.)
vapor pressure:
14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

grade

LR

vapor density

2.5 (vs air)

vapor pressure

14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

594 °F

expl. lim.

10.1 %

dilution

(for analytical testing)

refractive index

n20/D 1.385 (lit.)

pH

13 (20 °C, 100 g/L)

bp

55 °C (lit.)

mp

−50 °C (lit.)

density

0.707 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCNCC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Peyrton et al.
Molecules (Basel, Switzerland), 24(23) (2019-12-01)
Nowadays, polyols are basic chemicals for the synthesis of a large range of polymers, such as polyurethane foams (PUF), which are produced with several other compounds, such as polyisocyanates. During the last decades, the oleo-chemistry has developed several routes from
Katina S S Dossou et al.
Journal of chromatography. A, 1216(44), 7450-7455 (2009-06-26)
A polysaccharide-based chiral stationary phase (Sepapak-4), with cellulose tris(4-chloro-3-methylphenylcarbamate) as chiral selector, has been investigated in liquid chromatography (LC). Its enantioresolution power was evaluated towards 13 basic amino-drugs with widely different structures and polarities, using polar organic mobile phases. After
Roberto Cirilli et al.
Analytical chemistry, 81(9), 3560-3570 (2009-03-28)
Second-order rate constants of the diethylamine-promoted enantiomerization of 2-[2-(1-methyl-1H-pyrrol-2-yl)-2-oxo-1-phenylethyl]-isoindole-1,3-dione, a chiral alpha-substituted ketone endowed with high anti-MAO activity type-A, were measured by dynamic high-performance liquid chromatography (DHPLC), stopped-flow high-performance liquid chromatography (sf-HPLC), and a classical method based on enantioselective HPLC
Clarence M Ongkudon et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(28), 2719-2725 (2010-09-03)
Anion exchange monolithic chromatography is increasingly becoming a prominent tool for plasmid DNA purification but no generic protocol is available to purify all types of plasmid DNA. In this work, we established a simple framework and used it to specifically
Muhammad J A Shiddiky et al.
Journal of the American Chemical Society, 132(29), 10053-10063 (2010-07-06)
A highly selective and sensitive electrochemical biosensor has been developed that detects DNA hybridization by employing the electrocatalytic activity of ferrocene (Fc) bearing cyclen complexes (cyclen = 1,4,7,10-tetraazacyclododecane, Fc[Zn(cyclen)H(2)O](2)(ClO(4))(4) (R1), Fc(cyclen)(2) (R2), Fc[Zn(cyclen)H(2)O](ClO(4))(2) (R3), and Fc(cyclen) (R4)). A sandwich-type approach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service